ContactUsLinkComponent
CD45R/B220 Monoclonal Antibody, Clone: RA3-6B2, Host Species: Rat, Reactivity: Cat (Feline), Isotype: IgG2a, K, Conjugate: Spark YG 570, Immunogen: Abelson murine leukemia virus-induced pre-B tumor cells, Concentration: 5.0 - 10.0 ug/mL, Application: IHC-F, Size: 100uG
Catalog #: CA76522-712
Supplier:  Biolegend
CD45R/B220 Monoclonal Antibody, Clone: RA3-6B2, Host Species: Rat, Reactivity: Cat (Feline), Isotype: IgG2a, K, Conjugate: Spark YG 570, Immunogen: Abelson murine leukemia virus-induced pre-B tumor cells, Concentration: 5.0 - 10.0 ug/mL, Application: IHC-F, Size: 100uG
Catalog #: CA76522-712
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.