ContactUsLinkComponent
B Catenin 1 (CTNNB1) Monoclonal Antibody, Clone: 12F7, Host Species: Mouse, Reactivity: Human, Mouse, Rat, Isotype: IgG1, K, Conjugate: APC, Immunogen: Recombinant B-catenin (N-terminal fragment) fused to MBP, Application: ICFC, Size: 100 tests
Catalog #: CA76522-684
Supplier:  Biolegend
B Catenin 1 (CTNNB1) Monoclonal Antibody, Clone: 12F7, Host Species: Mouse, Reactivity: Human, Mouse, Rat, Isotype: IgG1, K, Conjugate: APC, Immunogen: Recombinant B-catenin (N-terminal fragment) fused to MBP, Application: ICFC, Size: 100 tests
Catalog #: CA76522-684
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.