ContactUsLinkComponent
CD5 Monoclonal Antibody, Clone: L17F12, Host Species: Mouse, Reactivity: Human, Isotype: IgG2a, K, Conjugate: PE/Cyanine5, Immunogen: Human T-acute lymphoblastic leukemia (ALL) cells, Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-676
Supplier:  Biolegend
CD5 Monoclonal Antibody, Clone: L17F12, Host Species: Mouse, Reactivity: Human, Isotype: IgG2a, K, Conjugate: PE/Cyanine5, Immunogen: Human T-acute lymphoblastic leukemia (ALL) cells, Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-676
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.