ContactUsLinkComponent
CD27 Monoclonal Antibody, Clone: M-T271, Host Species: Mouse, Reactivity: Baboon, Cynomolgus, Rhesus, Isotype: IgG1, K, Conjugate: PE/Cyanine5, Immunogen: Human T cells from a T-ALL patient, Application: FC, Size: 100 tests
Catalog #: CA76522-674
Supplier:  Biolegend
CD27 Monoclonal Antibody, Clone: M-T271, Host Species: Mouse, Reactivity: Baboon, Cynomolgus, Rhesus, Isotype: IgG1, K, Conjugate: PE/Cyanine5, Immunogen: Human T cells from a T-ALL patient, Application: FC, Size: 100 tests
Catalog #: CA76522-674
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.