ContactUsLinkComponent
HMOX1 Monoclonal Antibody, Clone: W19398C, Host Species: Rat, Reactivity: Human, Isotype: IgG2b, K, Conjugate: Purified, Concentration: 2.0 - 5.0 ug/mL, Application: WB, ICC, ICFC, IP, Size: 25uG
Catalog #: CA76522-592
Supplier:  Biolegend
HMOX1 Monoclonal Antibody, Clone: W19398C, Host Species: Rat, Reactivity: Human, Isotype: IgG2b, K, Conjugate: Purified, Concentration: 2.0 - 5.0 ug/mL, Application: WB, ICC, ICFC, IP, Size: 25uG
Catalog #: CA76522-592
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.