ContactUsLinkComponent
Buffer Set A5, includes Setup Beads, SA-PE, Assay buffer, Wash buffer, Plate Sealers, Matrix B5, Application: Multiplex - Component, Size: 100 tests
Catalog #: CA76522-562
Supplier:  Biolegend
Buffer Set A5, includes Setup Beads, SA-PE, Assay buffer, Wash buffer, Plate Sealers, Matrix B5, Application: Multiplex - Component, Size: 100 tests
Catalog #: CA76522-562
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.