ContactUsLinkComponent
CD161 Monoclonal Antibody, Clone: HP-3G10, Host Species: Mouse, Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Isotype: IgG1, K, Conjugate: PE/Cyanine5, Immunogen: Human NK cells, Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-560
Supplier:  Biolegend
CD161 Monoclonal Antibody, Clone: HP-3G10, Host Species: Mouse, Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Isotype: IgG1, K, Conjugate: PE/Cyanine5, Immunogen: Human NK cells, Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-560
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.