ContactUsLinkComponent
CD51 (Integrin AvB5) Monoclonal Antibody, Clone: P1F6, Host Species: Mouse, Reactivity: : Hamster, Isotype: IgG1, K, Conjugate: PE/Cyanine7, Immunogen: The P1F6 monoclonal antibody was generated against human small cell lung carcinoma cells (UCLA P3), Application: FC, Size: 100 tests
Catalog #: CA76522-546
Supplier:  Biolegend
CD51 (Integrin AvB5) Monoclonal Antibody, Clone: P1F6, Host Species: Mouse, Reactivity: : Hamster, Isotype: IgG1, K, Conjugate: PE/Cyanine7, Immunogen: The P1F6 monoclonal antibody was generated against human small cell lung carcinoma cells (UCLA P3), Application: FC, Size: 100 tests
Catalog #: CA76522-546
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.