ContactUsLinkComponent
IDH2 Monoclonal Antibody, Clone: KrMab-3, Host Species: Mouse, Reactivity: Human, Mouse, Isotype: IgG2b, K, Conjugate: Purified, Immunogen: Synthetic peptide corresponding to residues surrounding Ala174 of human IDH2 protein, Concentration: 2.0 - 5.0 ug/mL, Application: WB, ICC, IP, Size: 100uG
Catalog #: CA76522-486
Supplier:  Biolegend
IDH2 Monoclonal Antibody, Clone: KrMab-3, Host Species: Mouse, Reactivity: Human, Mouse, Isotype: IgG2b, K, Conjugate: Purified, Immunogen: Synthetic peptide corresponding to residues surrounding Ala174 of human IDH2 protein, Concentration: 2.0 - 5.0 ug/mL, Application: WB, ICC, IP, Size: 100uG
Catalog #: CA76522-486
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.