ContactUsLinkComponent
Cell-Vive T-NK Xeno-Free Serum Substitute, GMP, Formulation: Xeno-free, serum-free formulation, Storage & Handling: Store at -20 deg C to -5 deg C, Apps: Cell Culture, Size: 50ml
Catalog #: CA76522-462
Supplier:  Biolegend
Cell-Vive T-NK Xeno-Free Serum Substitute, GMP, Formulation: Xeno-free, serum-free formulation, Storage & Handling: Store at -20 deg C to -5 deg C, Apps: Cell Culture, Size: 50ml
Catalog #: CA76522-462
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.