ContactUsLinkComponent
CD88 (C5aR) Monoclonal Antibody, Clone: 20/70, Host Species: Rat, Reactivity: Mouse, Isotype: IgG2b, K, Conjugate: TotalSeq-B1042, Immunogen: mC5aR transfected cells, Concentration: 0.5 mg/mL, Application: PG, Size: 10uG
Catalog #: CA76522-434
Supplier:  Biolegend
CD88 (C5aR) Monoclonal Antibody, Clone: 20/70, Host Species: Rat, Reactivity: Mouse, Isotype: IgG2b, K, Conjugate: TotalSeq-B1042, Immunogen: mC5aR transfected cells, Concentration: 0.5 mg/mL, Application: PG, Size: 10uG
Catalog #: CA76522-434
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.