ContactUsLinkComponent
CD9 Monoclonal Antibody, Clone: 2A1/CD9, Host Species: Mouse, Reactivity: Rat, Isotype: IgG1, K, Conjugate: FITC, Concentration: 0.5 mg/mL, Application: Flow cytometry, Size: 25uG
Catalog #: CA76522-422
Supplier:  Biolegend
CD9 Monoclonal Antibody, Clone: 2A1/CD9, Host Species: Mouse, Reactivity: Rat, Isotype: IgG1, K, Conjugate: FITC, Concentration: 0.5 mg/mL, Application: Flow cytometry, Size: 25uG
Catalog #: CA76522-422
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.