ContactUsLinkComponent
TotalSeq-C0970 PE Streptavidin, Reactivity Human, Mouse, Rat, All Species, Formulation: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA, Concentration: 0.5 mg/mL, Application: PG, Size: 10uG
Catalog #: CA76522-340
Supplier:  Biolegend
TotalSeq-C0970 PE Streptavidin, Reactivity Human, Mouse, Rat, All Species, Formulation: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA, Concentration: 0.5 mg/mL, Application: PG, Size: 10uG
Catalog #: CA76522-340
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.