ContactUsLinkComponent
CD123 Monoclonal Antibody, Clone: S18016C, Host Species: Mouse, Reactivity: Human, Isotype: IgG1, K, Conjugate: APC, Immunogen: Hu CD123 transfectants, Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-304
Supplier:  Biolegend
CD123 Monoclonal Antibody, Clone: S18016C, Host Species: Mouse, Reactivity: Human, Isotype: IgG1, K, Conjugate: APC, Immunogen: Hu CD123 transfectants, Application: Flow cytometry, Size: 100 tests
Catalog #: CA76522-304
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.