ContactUsLinkComponent
I-A/I-E Monoclonal Antibody, Clone: M5/114.15.2, Host Species: Rat, Reactivity: Mouse, Isotype: IgG2b, K, Conjugate: Spark YG 581, Immunogen: Activated C57BL/6 mouse spleen cells, Concentration: 0.5 mg/mL, Application: Flow cytometry, Size: 100uG
Catalog #: CA76522-294
Supplier:  Biolegend
I-A/I-E Monoclonal Antibody, Clone: M5/114.15.2, Host Species: Rat, Reactivity: Mouse, Isotype: IgG2b, K, Conjugate: Spark YG 581, Immunogen: Activated C57BL/6 mouse spleen cells, Concentration: 0.5 mg/mL, Application: Flow cytometry, Size: 100uG
Catalog #: CA76522-294
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.