ContactUsLinkComponent
Recombinant Human ALPL (carrier-free), Source: Human ALPL, amino acid (Leu18 - Ser502) was expressed in CHO cells. The C-terminus contains 6His-tag. Molecular Mass: 491 amino acid recombinant protein has a predicted molecular mass of approximately 54.2 kD. Purity > 95%, Size: 100uG
Catalog #: CA76522-248
Supplier:  Biolegend
Recombinant Human ALPL (carrier-free), Source: Human ALPL, amino acid (Leu18 - Ser502) was expressed in CHO cells. The C-terminus contains 6His-tag. Molecular Mass: 491 amino acid recombinant protein has a predicted molecular mass of approximately 54.2 kD. Purity > 95%, Size: 100uG
Catalog #: CA76522-248
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.