ContactUsLinkComponent
GDNF Monoclonal Antibody, Clone: A16099G, Host Species: Mouse, Reactivity: Human, Isotype: IgG2b, K, Conjugate: GMP Ultra-LEAF Purified, Immunogen: Recombinant human GDNF, Concentration: 0.5 mg/mL, Apps: Neutralization, Size: 100uG
Catalog #: CA76522-236
Supplier:  Biolegend
GDNF Monoclonal Antibody, Clone: A16099G, Host Species: Mouse, Reactivity: Human, Isotype: IgG2b, K, Conjugate: GMP Ultra-LEAF Purified, Immunogen: Recombinant human GDNF, Concentration: 0.5 mg/mL, Apps: Neutralization, Size: 100uG
Catalog #: CA76522-236
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.