ContactUsLinkComponent
HDAC2 Monoclonal Antibody, Clone: 13G8C67, Host Species: Mouse, Reactivity: Human, Mouse, Isotype: IgG1, K, Conjugate: Purified, Immunogen: Human HDAC2 peptide (471-488 a.a.) conjugated to KLH, Concentration: 0.1 - 10 ug/ml, Application: WB, IP, ICC, KO/KD-WB, ChIP, Size: 25uG
Catalog #: CA76522-212
Supplier:  Biolegend
HDAC2 Monoclonal Antibody, Clone: 13G8C67, Host Species: Mouse, Reactivity: Human, Mouse, Isotype: IgG1, K, Conjugate: Purified, Immunogen: Human HDAC2 peptide (471-488 a.a.) conjugated to KLH, Concentration: 0.1 - 10 ug/ml, Application: WB, IP, ICC, KO/KD-WB, ChIP, Size: 25uG
Catalog #: CA76522-212
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.