ContactUsLinkComponent
CCL3 (MIP-1A) Monoclonal Antibody, Clone: W16009B, Host Species: Rat, Reactivity: Human, Isotype: IgG2a, K, Conjugate: Ultra-LEAF Purified, Immunogen: Recombinant Human CCL3, Apps: Neutralization, Size: 1mg
Catalog #: CA76522-194
Supplier:  Biolegend
CCL3 (MIP-1A) Monoclonal Antibody, Clone: W16009B, Host Species: Rat, Reactivity: Human, Isotype: IgG2a, K, Conjugate: Ultra-LEAF Purified, Immunogen: Recombinant Human CCL3, Apps: Neutralization, Size: 1mg
Catalog #: CA76522-194
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.