ContactUsLinkComponent
GMP Recombinant Human IL-21 (carrier-free), Source: Human IL-21, amino acid Gln30-Ser162, was expressed in E.coli. Molecular Mass: 133 amino acid recombinant protein has a predicted molecular mass of approximately 15.5 kD. Purity > 95%, Size: 25uG
Catalog #: CA76522-188
Supplier:  Biolegend
GMP Recombinant Human IL-21 (carrier-free), Source: Human IL-21, amino acid Gln30-Ser162, was expressed in E.coli. Molecular Mass: 133 amino acid recombinant protein has a predicted molecular mass of approximately 15.5 kD. Purity > 95%, Size: 25uG
Catalog #: CA76522-188
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.