ContactUsLinkComponent
Uvp Solo Elite, 6.3Mp Monochrome, EtBr emission filter pre-mounted on the Elite Mount, UVP Elite UV Transilluminator (302 nm), Resoultion: 3072 x 2048 pixel, from fluorescent DNA gels, fluorescent protein gels, and blue light gels, to colorimetry, Voltage: 115V
Catalog #: CA76522-040
Supplier:  Analytik Jena CA
Uvp Solo Elite, 6.3Mp Monochrome, EtBr emission filter pre-mounted on the Elite Mount, UVP Elite UV Transilluminator (302 nm), Resoultion: 3072 x 2048 pixel, from fluorescent DNA gels, fluorescent protein gels, and blue light gels, to colorimetry, Voltage: 115V
Catalog #: CA76522-040
Supplier:  Analytik Jena CA
Customers Who Bought This Also Bought