ContactUsLinkComponent
Ni Sepharose excel, 25 mL, a his-tagged protein purification resin for minimized Ni-leakage and maximized protein recovery when samples contain stripping agents
Catalog #: MPCA29832243
Supplier:  Cytiva
Ni Sepharose excel, 25 mL, a his-tagged protein purification resin for minimized Ni-leakage and maximized protein recovery when samples contain stripping agents
Catalog #: MPCA29832243
Supplier:  Cytiva
Customers Who Bought This Also Bought