ContactUsLinkComponent
respirator, NIOSH-certified N95 particulate filter, provides a minimum 95% filtration efficiency of 0.3 micron particles (though not oil resistant), has not been tested vs COVID-19 and can’t be recommended for efficacy or use, NOT for medical use. Size Small
Catalog #: 76521-766
Supplier:  Kimberly-Clark
respirator, NIOSH-certified N95 particulate filter, provides a minimum 95% filtration efficiency of 0.3 micron particles (though not oil resistant), has not been tested vs COVID-19 and can’t be recommended for efficacy or use, NOT for medical use. Size Small
respirator, NIOSH-certified N95 particulate filter, provides a minimum 95% filtration efficiency of 0.3 micron particles (though not oil resistant), has not been tested vs COVID-19 and can’t be recommended for efficacy or use, NOT for medical use. Size Small
Catalog #: 76521-766
Supplier:  Kimberly-Clark
Customers Who Bought This Also Bought