Medical Refrigerator & Freezer Battery Backup - Hardwire Silent Sentry System 2000 Watts 600 Amp Hours Modified Sine Wave
Catalog #: 76521-604
Supplier: MEDICANIX INC
Medical Refrigerator & Freezer Battery Backup - Hardwire Silent Sentry System 2000 Watts 600 Amp Hours Modified Sine Wave
Catalog #: 76521-604
Supplier: MEDICANIX INC
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
31,17 $ to 282,75 $
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
89,92 $ to 1 797,48 $
Nalgene® Self-Zeroing Buret Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
177,89 $ to 307,53 $
SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
146,85 $
Shimpo LCD Contact/Non-Contact Handheld Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
771,89 $