Medical Refrigerator & Freezer Battery Backup - Standalone Silent Sentry System 4000 Watts 600 Amp Hours Pure Sine Wave
Catalog #: 76521-592
Supplier: MEDICANIX INC
Medical Refrigerator & Freezer Battery Backup - Standalone Silent Sentry System 4000 Watts 600 Amp Hours Pure Sine Wave
Catalog #: 76521-592
Supplier: MEDICANIX INC
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
31,17 $ to 282,75 $
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
69,94 $
Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Supplier: Thermo Fisher Scientific
614,96 $ to 1 251,90 $
SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
146,85 $
Shimpo LCD Contact/Non-Contact Handheld Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
771,89 $