Medical Refrigerator & Freezer Battery Backup - Standalone Silent Sentry System 1200 Watts 100 Amp Hours Modified Sine Wave
Catalog #: 76521-562
Supplier: MEDICANIX INC
Medical Refrigerator & Freezer Battery Backup - Standalone Silent Sentry System 1200 Watts 100 Amp Hours Modified Sine Wave
Catalog #: 76521-562
Supplier: MEDICANIX INC
Customers Who Bought This Also Bought
Nalgene® Self-Zeroing Buret Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
$177.89 to $307.53
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
$89.92 to $1,797.48
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$31.17 to $282.75
Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Supplier: Thermo Fisher Scientific
$639.34 to $1,284.30
SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$146.85
Shimpo Non-Contact Pocket Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$368.65