Medical Refrigerator & Freezer Battery Backup - Mobile Reassurance System 3000 Watts 300 Amp Hours Modified Sine Wave
Catalog #: 76521-546
Supplier: MEDICANIX INC
Medical Refrigerator & Freezer Battery Backup - Mobile Reassurance System 3000 Watts 300 Amp Hours Modified Sine Wave
Catalog #: 76521-546
Supplier: MEDICANIX INC
Customers Who Bought This Also Bought
Nalgene® Self-Zeroing Buret Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
177,89 $ to 307,53 $
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
89,92 $ to 1 797,48 $
VWR® Single Channel Electronic Timer with Memory and Clock and Certificate of Calibration
Supplier: VWR International
75,03 $ to 739,18 $
Shimpo LCD Contact/Non-Contact Handheld Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
771,89 $