Medical Refrigerator & Freezer Battery Backup - Mobile Reassurance System 2000 Watts 600 Amp Hours Modified Sine Wave
Catalog #: 76521-540
Supplier: MEDICANIX INC
Medical Refrigerator & Freezer Battery Backup - Mobile Reassurance System 2000 Watts 600 Amp Hours Modified Sine Wave
Catalog #: 76521-540
Supplier: MEDICANIX INC
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
31,17 $ to 282,75 $
VWR® Single Channel Electronic Timer with Memory and Certificate of Calibration
Supplier: VWR International
77,73 $ to 1 082,63 $
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
48,82 $ to 442,63 $
Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Supplier: Thermo Fisher Scientific
639,34 $ to 1 284,30 $
Nalgene® Self-Zeroing Buret Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
177,89 $ to 307,53 $
VWR® Single Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration
Supplier: VWR International
49,22 $ to 492,14 $