Primer, Pr-1200, Red, one component, dilute solution of silane coupling agents that is used to increase the bonding of room temperature and heat curing silicones to a variety of surfaces such as glass, plastics, metals, ceramics, and masonry, Size: 2.9Kg
Catalog #: 76521-298
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.