Gun, Dual Cartridge Manual Dispensing, hand-held applicator that is used for dispensing Loctite adhesives in two component cartridges with a mix ratio of 1:1 or 2:1, Used for dispensing Loctite adhesives in dual cartridges, Size: 50Ml
Catalog #: 76521-308
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.