Bottle, Sika Primer 210T, Light Yellow, is a one component, general purpose, fast curing primer that is used to improve bonding of Sika products to a variety of substrates such as plastics, Flash Point: -4 deg C, Size: 250Ml
Catalog #: 76521-304
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.