Kit, 186 Silicone Elastomer Clear, is a two component, room temperature and heat curing, encapsulant that is used as a protective coating for electrical applications,provides flexibility, high tear strength, and high viscosity, 10:1 mix ratio, Size: 4Kg
Catalog #: 76521-302
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.