Kit, Mounting Stand, used with DYMAX 1200 & 2000 Flood Lamp UV Light Curing Systems for protecting the operator from UV light exposure when curing photosensitive adhesives, 3 sided shield that is used to block UV light, includes mounting hardware, pedestal, shield, work surface, and safety goggles
Catalog #: 76521-300
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.