Os-20 Silicone Fluid, Clear, is an odorless, non-VOC compound that is used for rinsing parts, replacing organic fluids, and cleaning contaminated surfaces, provides low toxicity, low surface tension, high purity, and evaporates at room temperature, Viscosity:0.8 at 25 C, Size: 15 Kg Pail
Catalog #: 76521-286
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.