Base, Rtv-3120, Red, easily mixed and poured, cures at room temperature, gives accurate reproduction of masters for mold making with primer, provides wide service temperature ranges and absorbs mechanical shock and vibration, Size: 4Kg
Catalog #: 76521-282
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.