Gloves, Chemical Resistant Nitrile, Fully Coated 26" (660Mm)Extended Gauntlet,Cotton Interlock Liner, Rough Finish, Royal Blue, Size: Large
Catalog #: CA76521-118
Supplier: Showa-Best Glove
Gloves, Chemical Resistant Nitrile, Fully Coated 26" (660Mm)Extended Gauntlet,Cotton Interlock Liner, Rough Finish, Royal Blue, Size: Large
Catalog #: CA76521-118
Supplier: Showa-Best Glove
Customers Who Bought This Also Bought
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
$15.73 to $345.18
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
$541.80
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$86.77 to $708.50
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$464.49