ContactUsLinkComponent
Gloves, Chemical Resistant, Unique "Hybrid", Flock-Lined, 100% Nitrile, 12" (305Mm), 11Mil, Rolled Cuff, Embossed, Blue, Size: XXL
Catalog #: CA76521-104
Supplier:  Showa-Best Glove
Gloves, Chemical Resistant, Unique "Hybrid", Flock-Lined, 100% Nitrile, 12" (305Mm), 11Mil, Rolled Cuff, Embossed, Blue, Size: XXL
Catalog #: CA76521-104
Supplier:  Showa-Best Glove
Customers Who Bought This Also Bought