Gloves, Chemical Resistant, Unique "Hybrid", Flock-Lined, 100% Nitrile, 12" (305Mm), 11Mil, Rolled Cuff, Embossed, Blue, Size: XS
Catalog #: CA76521-094
Supplier: Showa-Best Glove
Gloves, Chemical Resistant, Unique "Hybrid", Flock-Lined, 100% Nitrile, 12" (305Mm), 11Mil, Rolled Cuff, Embossed, Blue, Size: XS
Catalog #: CA76521-094
Supplier: Showa-Best Glove
Customers Who Bought This Also Bought
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
240,47 $ to 8 653,27 $
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
362,33 $
Nalgene® Cryoware Marker Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
142,56 $ to 406,56 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
464,49 $
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
534,61 $ to 1 523,31 $