ContactUsLinkComponent
Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 19" (483Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: XL
Catalog #: CA76521-070
Supplier:  Showa-Best Glove
Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 19" (483Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: XL
Catalog #: CA76521-070
Supplier:  Showa-Best Glove
Customers Who Bought This Also Bought