ContactUsLinkComponent
Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 15" (381Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: XL
Catalog #: CA76521-060
Supplier:  Showa-Best Glove
Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 15" (381Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: XL
Catalog #: CA76521-060
Supplier:  Showa-Best Glove
Customers Who Bought This Also Bought