Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 15" (381Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: Medium
Catalog #: CA76521-056
Supplier: Showa-Best Glove
Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 15" (381Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: Medium
Catalog #: CA76521-056
Supplier: Showa-Best Glove
Customers Who Bought This Also Bought
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
$1,747.88
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$86.77 to $708.50
Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Supplier: Thermo Fisher Scientific
$919.35 to $933.31
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$464.49
SP Bel-Art Mercury Collector, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$90.77 to $443.51