Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 15" (381Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: Small
Catalog #: CA76521-054
Supplier: Showa-Best Glove
Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 15" (381Mm), 22Mil, 6 Pr Per Polybag, 12 Polybags/Case, Bisque, Green, Size: Small
Catalog #: CA76521-054
Supplier: Showa-Best Glove
Customers Who Bought This Also Bought
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
$15.73 to $345.18
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
$541.80
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$271.77
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
$534.61 to $1,523.31