Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 13" (330Mm), 15Mil, Bisque Finish, Green, Size: Small
Catalog #: CA76521-038
Supplier: Showa-Best Glove
Gloves, Chemical Resistant Unsupported 100% Chlorinated Nitrile, Unlined, 13" (330Mm), 15Mil, Bisque Finish, Green, Size: Small
Catalog #: CA76521-038
Supplier: Showa-Best Glove
Customers Who Bought This Also Bought
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
541,80 $
Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Supplier: Thermo Fisher Scientific
1 263,71 $ to 1 282,92 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
271,77 $