ContactUsLinkComponent
Gloves, Biodegradable Single Use nitrile with EBT, powder-free, low-modulus 100%-nitrile, bisque fingertips, 9-1/2 in, 2.5-mil thickness, Color: blue, Size: Small
Catalog #: CA76521-000
Supplier:  Showa-Best Glove
Gloves, Biodegradable Single Use nitrile with EBT, powder-free, low-modulus 100%-nitrile, bisque fingertips, 9-1/2 in, 2.5-mil thickness, Color: blue, Size: Small
Catalog #: CA76521-000
Supplier:  Showa-Best Glove
Customers Who Bought This Also Bought