ContactUsLinkComponent
Filter Plate, AcroPrep Advance 96-Well, 1 mL, 30-40 um PP/PE non-woven media, can be used in a number of aqueous high throughput filtration applications including gross fractionation, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA30621-080
Supplier:  Cytiva
Filter Plate, AcroPrep Advance 96-Well, 1 mL, 30-40 um PP/PE non-woven media, can be used in a number of aqueous high throughput filtration applications including gross fractionation, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA30621-080
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.