Smart-Vue Pro temp sensor for high temp range (+100C to +1100C). This sensor outputs to a 4-20mA connection. This must be purchased in conjunction with 4-20mA sensor (SVPSEN4CS001). Not calibrated.
Catalog #: 76520-816
Supplier: Thermo Fisher Scientific
Smart-Vue Pro temp sensor for high temp range (+100C to +1100C). This sensor outputs to a 4-20mA connection. This must be purchased in conjunction with 4-20mA sensor (SVPSEN4CS001). Not calibrated.
Catalog #: 76520-816
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$31.17 to $282.75
VWR® Single Channel Electronic Timer with Memory and Certificate of Calibration
Supplier: VWR International
$77.73 to $1,082.63
Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Supplier: Thermo Fisher Scientific
$576.51 to $1,145.47
SP Bel-Art Three-In-One Multi-Channel Pipette Reservoir, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$146.85