Smart-Differential pressure sensor, for use with Smart-Vue Pro systems. Range: -500 Pa to +500 Pa. (-2 to +2 in./-5 to +5 cm in H2O) Powered by the Smart-Vue Pro module. Tube length: 50 cm/19.7 in. Diameter: 4 mm/1.9 in. (4-5 mm/1.9-2 in. Adapter). Not calibrated.
Catalog #: 76520-804
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought