Smart-CO2 sensor, with temp and humidity sensors for use with Smart-Vue Pro systems. CO2 range: 0.5% to 9.9%. Temp range: 0 deg C to +50 deg C, Relative humidity range: 0% to 99%. 5V external power supply. Not calibrated.
Catalog #: 76520-800
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought