ContactUsLinkComponent
Filter Plate, AcroPrep Advance, for DNA Purification - 1 mL, DNA binding, ensures high yields of contaminant-free DNA for downstream applications, such as PCR and sequencing, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA97052-134
Supplier:  Cytiva
Filter Plate, AcroPrep Advance, for DNA Purification - 1 mL, DNA binding, ensures high yields of contaminant-free DNA for downstream applications, such as PCR and sequencing, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA97052-134
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.