ContactUsLinkComponent
Filter Plate, AcroPrep Advance 96-Well, 1 mL, 1.2 um Supor membrane, can be used in a number of high throughput applications including aqueous filtration, protein purification and multiplex assays, Height (Metriccm)1.8cm, Length: 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene
Catalog #: MPCA97052-128
Supplier:  Cytiva
Filter Plate, AcroPrep Advance 96-Well, 1 mL, 1.2 um Supor membrane, can be used in a number of high throughput applications including aqueous filtration, protein purification and multiplex assays, Height (Metriccm)1.8cm, Length: 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene
Catalog #: MPCA97052-128
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.